8-Hydroxy-2'-deoxyguanosine ≥98%

Supplier: Enzo Life Sciences

ALEXALX480090M001EA 126 EUR
ALEXALX480090M001
8-Hydroxy-2'-deoxyguanosine ≥98%
8-Hydroxy-2'-deoxyguanosine

Oxidative DNA degradation product.


Soluble in water (15 mg/ml) or DMSO (12 mg/ml).

Formula: C₁₀H₁₃N₅O₅
MW: 283,244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788705
CAS Number: 88847-89-6

Order Now

Specification Test Results

Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR