15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid ≥95% (by HPLC)

Supplier: Enzo Life Sciences
Danger

ALEXBML-K015-0050EA 279 EUR
ALEXBML-K015-0050
15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid ≥95% (by HPLC)
15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid

Biosynthesis via 15-OH prostaglandin dehydrogenase from lung and liver microsomes and by a nonenzymatic process.

Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 81416-72-0
UN: 1170
ADR: 3,II
https://export.vwr.com/stibo/web/std.lang.all/13/08/45401308.jpg

Order Now

Specification Test Results

Purity ≥95% (HPLC)
Concentration (for 50 µg size) 50 µg/ml for 50 µg size
Concentration (for 1 mg size) 1 mg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR