L(+)-Aspartic acid, SAFC®
Supplier: Merck
M-Clarity™ quality level = MQ400
SIALA5474-1KGEA
365
EUR
SIALA5474-1KG
L(+)-Aspartic acid, SAFC®
L(+)-Aspartic acid
Formula:
C₄H₇NO₄ MW: 133,1 g/mol Boiling Pt: 311 °C (1013 hPa) Melting Pt: 251 °C Density: 1,6613 g/cm³ (13 °C) |
MDL Number:
MFCD00002616 CAS Number: 56-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...