Ponasterone A ≥95% (by HPLC)

Supplier: Enzo Life Sciences

ALEXALX370014M001EA 89.6 EUR
ALEXALX370014M001 ALEXALX370014M005 ALEXALX370014M100
Ponasterone A ≥95% (by HPLC)
Ponasterone A

Ecdysteroid.


A member of the ecdysteroid family. Analog of ecdysone with similar properties to muristerone A. It is a functional, reliable and economical substitute for muristerone A as an inducer for the ecdysone-inducible mammalian expression system. Results show that ponasterone A was able to induce expression of β-galactosidase to levels similar to those obtained with muristerone A induction. Potent compound for ecdysteroid activity; EC₅₀ = 0.07 µM.


Soluble in acetic acid, 100% ethanol, methanol or DMSO; insoluble in water.

Formula: C₂₇H₄₄O₆
Storage Temperature: Freezer
CAS Number: 13408-56-5

Order Now

Specification Test Results

Purity ≥95% (HPLC)
Identity Identity determined by ¹H- and ¹³C-NMR
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR