NS 1738 ≥99% (by HPLC)
Supplier: Enzo Life Sciences
ALEXENZCHM1170010EA
220
EUR
ALEXENZCHM1170010
ALEXENZCHM1170050
NS 1738 ≥99% (by HPLC)
NS 1738
A selective positive allosteric modulator selective for α7 nicotinic acetylcholine receptors (α7 nAChR).
NS 1738 exhibits cognitive enhancing effects in vivo.
Soluble in DMSO or ethanol.
Formula:
C₁₄H₉Cl₂F₃N₂O₂ MW: 365,1 g/mol Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
MDL Number:
MFCD11100189 CAS Number: 501684-93-1 |
Specification Test Results
Purity | ≥99% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...