Dibromochloromethane

48542.EA 112 EUR
48542.
Dibromochloromethane
Dibromochloromethane
Formula: Br₂ClCH
MW: 208,28 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: –22 °C
Density: 2,445 g/cm³ (15 °C)
MDL Number: MFCD00000820
CAS Number: 124-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR