Lysates, rat brain
Supplier: Enzo Life Sciences
ALEXBML-SW103-0200EA 54.6 EUR
ALEXBML-SW103-0200
Lysates, rat brain
Cells
Whole brain protein extract from rat is useful as a Western blotting control for anti-nNOS and PKA RIIβ antibody or for use with other antibodies.
  • Ready to use
  • Use 5 µl/lane
Delivery information: Supplied as tissue extract (approximately 5 mg/ml) in SDS-PAGE sample buffer.