Urea

EDQMU0600000EA 235 EUR
EDQMU0600000
Urea
Urea
Formula: NH₂CONH₂
MW: 60,06 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 132,7 °C
Density: 1,34 g/cm³ (20 °C)
MDL Number: MFCD00008022
CAS Number: 57-13-6
Merck Index: 12,10005
https://export.vwr.com/stibo/web/std.lang.all/70/46/2297046.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR