Data analysis software, Assay Blaster
Supplier: Enzo Life Sciences
ALEXADI-28-0002EA 147 EUR
ALEXADI-28-0002
Data analysis software, Assay Blaster
Software Laboratory Software
Assay blaster data analysis software offers a simple and cost-effective solution for analysing assay results.
  • Simplified data analysis
  • Parameters and settings can be manually adjusted
  • Optionally use duplicate or triplicate data series and remove outliers
  • Fully configurable custom report generation
It can be used to analyse mean signal, net signal, or %B/Bo results and extrapolate concentration values of unknown samples using point-to-point, 4PL, or 5PL curve fit methods. The intuitive user interface makes getting started easy.