D-erythro-C8-ceramine ≥98% (by TLC)

Supplier: Enzo Life Sciences

ALEXBML-SL145-0001EA 96.6 EUR
ALEXBML-SL145-0001 ALEXBML-SL145-0005
D-erythro-C8-ceramine ≥98% (by TLC)
D-erythro-C8-ceramine

Ceramide-activated protein kinase activator.


C₈ ceramine is an analog of ceramide in which the amide is substituted with an amine. This substitution renders it inert to ceramidases. It is highly effective at induction of apoptosis in U937 cells. Ceramine induces peak DNA fragmentation in 6 hours compared to 18 hours for C₈ ceramide.


Soluble in ethanol (25 mg/ml) or DMSO (25 mg/ml).

Formula: C₂₆H₅₃NO₂
MW: 411,715 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01632747
CAS Number: 170926-06-4

Order Now

Specification Test Results

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR