Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥85% (by HPLC) ATP analog
Supplier: Enzo Life Sciences
|
Warning
|
ALEXBML-ST101-0025EA
615
EUR
ALEXBML-ST101-0025
ALEXBML-ST101-0005
Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥85% (by HPLC) ATP analog
Adenosine-5'-O-(3-thiotriphosphate) tetralithium salt
ATP analog.
An ATP analog that can substitute for ATP in various kinase reactions. The resulting thiophosphorylated proteins are resistant to dephosphorylation by protein phosphatases.
Soluble in water (25 mg/ml).
Formula:
C₁₀H₁₂N₅O₁₂PS·4Li MW: 546,98 g/mol Storage Temperature: Freezer |
CAS Number:
93839-89-5 |
Specification Test Results
Purity | ≥85% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...