Lipopolysaccharides (from S. abortus equi) S-form, TLRgrade™ ready-to-use

Supplier: Enzo Life Sciences
Danger

ALEXALX581150-R500EA 573 EUR
ALEXALX581150-R500
Lipopolysaccharides (from S. abortus equi) S-form, TLRgrade™ ready-to-use
Lipopolysaccharides

TLR4 activator.


Smooth (S)-form LPS, isolated and purified from Salmonella abortus equi by modification of the phenol water extraction and PCP method. LPS-Biotin was prepared using the biotin reagent biotinamidocaproate N-hydroxysuccinimide ester.


Useful reagent for evaluating LPS binding and cell activation in white blood cells.

Order Now

Specification Test Results

Concentration 1 mg/ml
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double-distilled water
Purity Absence of detectable protein or DNA contaminants with agonistic TLR activity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR