MPLA (from E.coli) ≥99.9% serotype R515 (Re), TLRgrade™ ready-to-use
Supplier: Enzo Life Sciences
ALEXALX581203R500EA
495
EUR
ALEXALX581203R500
ALEXALX581203L001
MPLA (from E.coli) ≥99.9% serotype R515 (Re), TLRgrade™ ready-to-use
MPLA
TLR4 activator.
Isolated from E. coli, Serotype R515 (Re mutant). Prepared by acid hydrolysis from purified E. coli, Serotype R515 (Re) LPS (isolated by a modification of the PCP extraction method), converted to the uniform salt form.
Activates TLR4 (Toll-like receptor 4). Does not activate TLR2 as determined with splenocytes and macrophages from TLR4 deficient mice. Exhibits reduced endotoxic and pyrogenic activity compared to lipid A and LPS.
Specification Test Results
Concentration | 1 mg/ml |
Formulation | Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water |
Purity | ≥99.9% (regarding protein or DNA contaminants, KDO not detectable) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...