Guanfacine hydrochloride ≥99% (by TLC)

Supplier: Enzo Life Sciences
Danger

ALEXBML-AR102-0100EA 108 EUR
ALEXBML-AR102-0100 ALEXBML-AR102-0500
Guanfacine hydrochloride ≥99% (by TLC)
Guanfacine hydrochloride

Adrenergic receptor ligand.


Centrally acting α2-adrenergic receptor agonist. In the guinea pig cerebral cortex, the Kd is 20 and 880 nM for the α2A- and the α2C-adrenoceptor, respectively. Antihypertensive agent.


Soluble in water (25 mg/ml).

Formula: C₉H₉Cl₂N₃O
MW: 282,55 g/mol
MDL Number: MFCD00798230
CAS Number: 29110-48-3
UN: 2811
ADR: 6.1,III
https://export.vwr.com/stibo/web/std.lang.all/34/57/45403457.jpg

Order Now

Specification Test Results

Purity ≥99% (HPLC)
Identity Determined by NMR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR