Clothiapine ≥99%
Supplier: Enzo Life Sciences
|
Danger
|
ALEXBML-D108-0100EA
223
EUR
ALEXBML-D108-0100
ALEXBML-D108-0020
Clothiapine ≥99%
Clothiapine
Dopamine receptor ligand.
A typical neuroleptic agent related to clozapine.
Soluble in DMSO (10 mg/ml).
Formula:
C₁₈H₁₈ClN₃S |
CAS Number:
2058-52-8 EINECS: 218-162-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity | ≥99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...