Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Sigma-Aldrich®
Supplier: ROCHE
Synonyms:
Tris (Hydroxymethyl) Aminomethane
, Trometamol
, 2-Amino-2-(hydroxymethyl)propane-1,3-diol
M-Clarity™ quality level = MQ100
ROCH10708976001EA
192
EUR
ROCH10708976001
ROCH708976
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Sigma-Aldrich®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula:
NH₂C(CH₂OH)₃ MW: 121,14 g/mol Boiling Pt: 357 °C (1013 hPa) Melting Pt: 169…172 °C Density: 1,328 g/cm³ (20 °C) |
MDL Number:
MFCD00004679 CAS Number: 77-86-1 Merck Index: 12,09902 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...