Vinyl acetate
48486.EA
51.2
EUR
48486.
Vinyl acetate
Vinyl acetate
Formula:
C₄H₆O₂ MW: 86,09 g/mol Boiling Pt: 72,3 °C (1013 hPa) Melting Pt: –100 °C Density: 0,932 g/cm³ (20 °C) |
MDL Number:
MFCD00008713 UN: 1301 ADR: 3,II Merck Index: 12,10130 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...