4-Diphenylacetoxy-N-methylpiperidine methiodide ≥98% (by HPLC)

Supplier: Enzo Life Sciences

ALEXBML-C104-0050EA 286 EUR
ALEXBML-C104-0050
4-Diphenylacetoxy-N-methylpiperidine methiodide ≥98% (by HPLC)
4-Diphenylacetoxy-N-methylpiperidine methiodide

Acetylcholine receptor ligand.


M₃ muscarinic receptor antagonist.


Soluble in DMSO and warm water.

Formula: C₂₁H₂₆INO₂
MW: 451,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078564
CAS Number: 1952-15-4
https://export.vwr.com/stibo/web/std.lang.all/34/62/45403462.jpg

Order Now

Specification Test Results

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR