Barium diphenylamine-4-sulphonate, TECHNICAL

Supplier: VWR Chemicals

84003.100EA 138 EUR
84003.100
Barium diphenylamine-4-sulphonate, TECHNICAL
Barium diphenylamine-4-sulphonate
Formula: C₂₄H₂₀BaN₂O₆S₂
MW: 633,89 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00007497
CAS Number: 6211-24-1
UN: 1564
ADR: 6.1,III
https://export.vwr.com/stibo/web/std.lang.all/76/37/2297637.jpg

Order Now

Specification Test Results

IR Spectrum Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR