4-Nitrophenol

Supplier: Merck

48549.EA 44.8 EUR
48549.
4-Nitrophenol
4-Nitrophenol
Formula: C₆H₅NO₃
MW: 139,11 g/mol
Boiling Pt: 279 °C (1013 hPa)
Melting Pt: 110…114 °C
Density: 1,48 g/cm³ (20 °C)
Flash Pt: 169 °C
MDL Number: MFCD00007331
UN: 1663
ADR: 6.1,III
Merck Index: 12,06718

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR