4-Chloro-m-cresol

Supplier: Merck

SIAL24940-1KG-FEA 82 EUR
SIAL24940-1KG-F
4-Chloro-m-cresol
4-Chloro-m-cresol
Formula: C₇H₇ClO
MW: 142,58 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 63…65 °C
Density: 0,9 g/cm³ (20 °C)
Flash Pt: 118 °C
MDL Number: MFCD00002323
UN: 3437
ADR: 6.1,II
Merck Index: 13,02152

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR