o-Toluidine, Supelco®
M-Clarity™ quality level = MQ100
45979-250MGEA
16.2
EUR
45979-250MG
o-Toluidine, Supelco®
o-Toluidine
Formula:
C₇H₉N MW: 107,16 g/mol Boiling Pt: 200 °C (1013 hPa) Melting Pt: < –15 °C Density: 0,998 g/cm³ (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00007730 UN: 1708 ADR: 6.1,II Merck Index: 12,09674 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...