2,4-Diaminotoluene analytical standard, Supelco®
442311.EA
22.9
EUR
442311.
2,4-Diaminotoluene analytical standard, Supelco®
2,4-Diaminotoluene
Formula:
C₇H₁₀N₂ MW: 122,17 g/mol Boiling Pt: 292 °C (1013 hPa) Melting Pt: 99 °C Density: 1,045 g/cm³ (20 °C) |
MDL Number:
MFCD00007804 UN: 1709 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...