Water, BioXtra, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIALW1503-500MLEA
74.6
EUR
SIALW1503-500ML
SIALW1503-100ML
Water, BioXtra, Sigma-Aldrich®
Water
Water for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Formula:
H₂O MW: 18,02 g/mol Boiling Pt: 100 °C (1013 hPa) Melting Pt: 0 °C Density: 1 g/cm³ (4 °C) |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 Merck Index: 12,10175 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...