Lead selenide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL254231-25GEA 438 EUR
SIAL254231-25G
Lead selenide, Sigma-Aldrich®
Lead selenide
Formula: PbSe
MW: 286,16 g/mol
Melting Pt: 79,5…80 °C
Density: 8,1 g/cm³ (20 °C)
MDL Number: MFCD00016273
UN: 3283
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR