Caps for tissue culture jars, B-cap, Magenta™
Supplier: Merck
SIALB8648-100EAEA 184 EUR
SIALB8648-100EA SIALB8648-500EA
Caps for tissue culture jars, B-cap, Magenta™
Closures Bottle Closures
Designed for use in plant tissue culture laboratories using baby food culture jars (V8630 and V0633).
  • Autoclavable
The unique design facilitates excellent light penetration and easy usage.