C1-C6 n-Paraffins, each component in nitrogen, analytical standards, Supelco®
Supplier: Merck
501859.EA 408 EUR
501859. 23444. 330300. 501786. 501840. 501778.
C1-C6 n-Paraffins, each component in nitrogen, analytical standards, Supelco®
Standards Petroleum Industry Standards
Multi-component gas mixture suitable for GC calibration.
  • A certificate of analysis is included.
  • Packaged in lightweight, non-refillable, recyclable containers.
  • Portable, convenient, and easy to use.
  • Manufactured gravimetrically for maximum accuracy.
  • NIST traceable.