Organic reference standard, Bezafibrate, analytical standard, Supelco®
Supplier: Merck
72516-500MGEA 60.2 EUR
72516-500MG
Organic reference standard, Bezafibrate, analytical standard, Supelco®
Standards Organic Standards
The peroxisome proliferator-activated receptor (PPAR) is a member of the steroid nuclear receptor superfamily. Bezafibrate is a peroxisome proliferator-activated receptor agonist for PPARα, PPARδ, and PPARγ. Lipoprotein lipase (LPL) activator.
PPARgamma agonists, including Bezafibrate, have beneficial effects in the suppression of the inflammatory response during RSV infection and therefore might have clinical efficacy in the course of severe RSV-infection.