Pesticide reference standard, Simeton, PESTANAL®, analytical standard, Supelco®
Supplier: Merck
PESTANAL®
32072-25MGEA 163 EUR
32072-25MG
Pesticide reference standard, Simeton, PESTANAL®, analytical standard, Supelco®
Standards Organic Standards
Simeton has been used as internal standard in ESI-LC/MS method for quantifying environmental contaminants and in LC-MS/MS analysis for determination of pesticide residue from constructed wetlands.
  • It is an alkoxytriazine mostly used as pesticide