Aluminium trilactate, TECHNICAL

83861.230EA 17 EUR
83861.230
Aluminium trilactate, TECHNICAL
Aluminium trilactate
Formula: (H₃CCH(OH)COO)₃Al
MW: 294,19 g/mol
Melting Pt: > 300 °C
MDL Number: MFCD00043086
CAS Number: 18917-91-4
Merck Index: 13,00347

Order Now

Specification Test Results

Assay (calculated as Al) Min. 8.6 %
Assay (on anhydrous substance) Min. 94 %
Heavy metals (as Pb) Max. 20 ppm
Water Max. 5 %
Cl (Chloride) Max. 0.04 %
SO4 (Sulphate) Max. 0.6 %
Fe (Iron) Max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR