Anti-Streptavidin Antibody (HRP (Horseradish Peroxidase))

Supplier: Cytiva
RPN4401EA 1133 EUR
RPN4401
Anti-Streptavidin Antibody (HRP (Horseradish Peroxidase))
Antibodies
For use in ELISA systems. For ELISA, it is recommended that the conjugate should be diluted in the range 1:4000 to 1:8000.

Amdex conjugates are already supplied as components of the Biotrak high-sensitivity cytokine ELISAs, complete with fully-optimized protocols. Easily soluble in cold and hot water.

Type:
Antigen: Streptavidin
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR