MOPS-Na ≥99%, Proteomics Grade

M215-100GEA 88 EUR
M215-100G
MOPS-Na ≥99%, Proteomics Grade
MOPS-Na

MOPS is a zwitterionic buffer used as a running buffer for denaturing agarose gel electrophoresis of RNA. Having a buffering range from 6.5 - 7.9, MOPS works exceptionally well with formaldehyde gels at 20 mM concentration.

Formula: C₇H₁₄NNaO₄S
MW: 231,25 g/mol
MDL Number: MFCD00064350
CAS Number: 71119-22-7

Order Now

Specification Test Results

Abs.@260 nm (0.1 M, Water) <0.03
DNase none detected
Heavy Metals <0.005%
Appearance White powder
Loss on drying <1%
pH (0.1 M, Water) @25 °C 10 − 11
pKa (20 °C) 7 − 7.4
Protease none detected
Purity ≥99%
RNase none
Solubility (0.1 M, Water) Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR