PVP-Free polycarbonate cell culture membrane, Whatman™
Supplier: Whatman products (Cytiva)
Whatman™
WHAT10418101EA 81.7 EUR
WHAT10418101
PVP-Free polycarbonate cell culture membrane, Whatman™
Filters Membrane Filters
Cytiva's Whatman cell culture membrane filters are made with PVP-free polycarbonate for enhanced cellular adhesion in biological applications such as neutrophil chemotaxis.
  • Cell culture filter suitable for biological applications
  • Thin and uniform cylindrical pores for rapid cell migration
  • Offered without the standard PVP wetting agent for increased cellular adhesion in applications such as neutrophil chemotaxis
  • Reduces incubation time and the need to sterilize
Effective chemotaxis requires a delicate balance between cell adhesion and migration. This Whatman Nuclepore membrane filter from Cytiva's Life Sciences business combines PVP-free polypropylene with sharp, uniform cylindrical pores to achieve both.