Labels, thread marking cards with digital legend, type B-500
Supplier: Brady
BRDY010011EA 61.72 EUR
BRDY010011 BRDY010001 BRDY010012 BRDY010010 BRDY010008 BRDY010009 BRDY010006 BRDY010007 BRDY010004 BRDY010005 BRDY010002 BRDY010003
Labels, thread marking cards with digital legend, type B-500
Brady B­500 is a polymer coated cloth with a printable topcoat and a rubber based pressure sensitive adhesive.
  • Good oil and water resistance
  • Good print durability
  • Excellent flexibility for wrapping around curved surfaces
This a general purpose material for a variety of pre­printed labeling and wire marking applications requiring durability and economy.