Tris-Glycine-SDS 10X Running Buffer
Supplier: Apollo
APOSBI2078-5PACKSEA 0 EUR
APOSBI2078-5PACKS APOSBI2078-10PACKS
Tris-Glycine-SDS 10X Running Buffer
Buffers Electrophoresis Buffers Running Buffers
Pre-weighed sachet containing sufficient buffer salts to create 1 litre of solution containing 1M TRIS, 1M HEPES and 30mM SDS.