Phenol stabilised and with separate Tris buffer solution

Danger

APOSBIA0447-500MLEA 0 EUR
APOSBIA0447-500ML
Phenol stabilised and with separate Tris buffer solution
Phenol

Stabilised with 0.1 % 8-Hydroxyquinoline; with separate Tris buffer (A2492).


Shelf life approx. 15 months.

Formula: C₆H₅OH
MW: 94,11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00002143
UN: 2821
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR