Water, Molecular Biology Grade

Supplier: Apollo Scientific

APOSBIA7398-1LEA 72.7 EUR
APOSBIA7398-1L APOSBIA7398-500ML
Water, Molecular Biology Grade
Water
Formula: H₂O
MW: 18,02 g/mol
Boiling Pt: 100 °C
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR