Calyculin A

Danger

APOSBIC1019-100UGEA 0 EUR
APOSBIC1019-100UG APOSBIC1019-10UG APOSBIC1019-50UG APOSBIC1019-300UG APOSBIC1019-100MG APOSBIC1019-10MG APOSBIC1019-50MG
Calyculin A
Calyculin A

PP1 inhibitor, PP2 inhibitor, serine/threonine phosphatatase inhibitor.

Formula: C₅₀H₈₁N₄O₁₅P
Storage Temperature: Freezer
CAS Number: 101932-71-2
UN: 3462
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR