Urea 99.5+%, Molecular Biology Grade
Supplier: Apollo Scientific
|
Warning
|
APOSBIU4110-1KGEA
72.7
EUR
APOSBIU4110-1KG
APOSBIU4110-5KG
Urea 99.5+%, Molecular Biology Grade
Urea
Chaotropic agent used in denaturing polyacrylamide gel electrophoresisDNases/RNases/Proteases not detected
- Protein
Formula:
NH₂CONH₂ MW: 60,06 g/mol Melting Pt: 132…135 °C Density: 1,335 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008022 CAS Number: 57-13-6 EINECS: 200-315-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...