Cryogenic tape for metal racks, Cryo-TAPE™
Supplier: GA International
817-0484EA 33.2 EUR
817-0484 817-0483 817-0473 817-0485 817-0480 817-0478 GAINTWA-25C1-50RE 817-0482 817-0470 817-0476 817-0481 817-0472 817-0471 817-0477 817-0486 817-0475 817-0479 817-0474
Cryogenic tape for metal racks, Cryo-TAPE™
Tapes
Cryogenic tape for the identification of various metal containers, for long-term cryogenic storage, as low as –196 °C (–321 °F).
  • Designed for use in harsh cryogenic conditions
  • Use on metal surfaces, including stainless steel and aluminium
  • Available in a variety of colours for easy colour coding and identification
  • Continuous tape format allows use of as much or little as necessary
Cryo-TAPE™ cryogenic tape of core size 25,4 mm, for metal racks can be used to identify stainless-steel and aluminium racks, X-ray cassettes, film cans, and more. It should be applied at room temperature before being placed in liquid nitrogen or ultra-low temperature freezers for subsequent storage. Writable with permanent cryo markers, the tape for metal racks can withstand storage in liquid nitrogen, transportation on dry ice, multiple freeze-thaw cycles, as well as gamma irradiation (up to 50 KGy).