Masterflex® Centrifuge Bottles, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
MFLX06059-92EA 253.9 EUR
MFLX06059-92
Masterflex® Centrifuge Bottles, Avantor®
Bottles Centrifuge Bottles
Process cell suspensions and lysates or tissue homogenates.
  • Feature high-visibility graduations, frosted label area, and flat bottom
  • Reuse and autoclave as needed—thick walls can handle repeated use
  • Available in polycarbonate or polypropylene
  • Choose screw caps for low-speed or sealing caps for high-speed applications
polypropylene (PP) cap.