Magnetic separation rack
Supplier: BioVision
M1501-1EA 367 EUR
M1501-1
Magnetic separation rack
Racks Magnetic Separation Racks
A rack to separate magnetic beads in tubes.
BioVision’s Magnetic Separation Rack is used to capture and separate magnetic beads used in magnetic beads based purification techniques. The rack captures magnetic beads which may be bound to DNA, viruses or cells in tubes. The biomolecules or cells are eluted and concentrated from the magnetic beads using appropriate buffers for downstream applications. BioVision’s Magnetic Rack uses permanent magnets and has a compact design to accommodate different sizes of tubes including twelve 1.5 ml tubes, two 15 ml tubes and two 50 ml tubes.