TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 98%

BLDPBD17161-500GEA 57 EUR
BLDPBD17161-500G BLDPBD17161-100G BLDPBD17161-5G BLDPBD17161-1000G BLDPBD17161-25G
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) 98%
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)

Storage condition: Inert atmosphere, Room Temperature

Formula: NH₂C(CH₂OH)₃·HCl
MW: 157,6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR