Acetic acid glacial

Supplier: U.S. Pharmacopeia

USPH1005706EA 471 EUR
USPH1005706
Acetic acid glacial
Acetic acid
Formula: H₃CCOOH
MW: 60,05 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 38,5 °C
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2789
ADR: 8,II
https://export.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR