Acetic acid ≥99.4%, Environmental Grade Plus

Supplier: Thermo Scientific

38739.APEA 535 EUR
38739.AP 38739.AK
Acetic acid ≥99.4%, Environmental Grade Plus
Acetic acid

CAS No.: 64-19-7
Molecular Formula: CH3CO2H
Formula Weight: 60.05
Flash Point: 40°(104°F)
Physical Form: Liquid
Density: 1.049
Refractive Index: 1.3721
Odour: Pungent
pH: 2,5
Melting Point: 16.6°
Boiling Point: 118.1°
Merck Reference: 14,55
UN No.: 2789
MDL No.: MFCD00036152

Formula: H₃CCOOH
MW: 60,05 g/mol
Boiling Pt: 118,1 °C
Melting Pt: 16,6 °C
Density: 1,049
Flash Pt: 40 °C (104 °F)
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2789
ADR: 8,II
https://export.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR