Sulphuric acid 0.005 mol concentrated aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
4869.EA
0
EUR
4869.
Sulphuric acid 0.005 mol concentrated aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sulphuric acid
Formula:
H₂SO₄ MW: 98,08 g/mol Density: 1,01 g/cm³ (20 °C) |
MDL Number:
MFCD00064589 UN: 2796 ADR: 8,II |
Specification Test Results
Titer (N) | 0.0099-0.0101 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...