X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Supplier: ROCHE
M-Clarity™ quality level = MQ100
ROCH11680293001EA
152
EUR
ROCH1680293
ROCH11680293001
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formula:
C₁₄H₁₅BrClNO₆ MW: 408,63 g/mol Melting Pt: 237…239 °C |
MDL Number:
MFCD00005666 CAS Number: 7240-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...