N-Octyl-β-D-glucopyranoside ≥98%, Reagent Grade

0479-5GEA 223 EUR
0479-5G
N-Octyl-β-D-glucopyranoside ≥98%, Reagent Grade
N-Octyl-β-D-glucopyranoside

Non ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.


These non-denaturing, gentler detergents are ideal for studying the conformatity.

Formula: C₁₄H₂₈O₆
MW: 292,37 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00063288
CAS Number: 29836-26-8

Order Now

Specification Test Results

Protease none detected
Purity ≥98%
Solubility (10%, Water) Pass
Specific rotation From −32 to −28°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR