Endosulfan (α- and β-isomers) ≥98%, PESTANAL® analytical reagent, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

32015-250MGEA 58.6 EUR
32015-250MG
Endosulfan (α- and β-isomers) ≥98%, PESTANAL® analytical reagent, Supelco®
Endosulfan (α- and β-isomers)
Formula: C₉H₆Cl₆O₃S
MW: 406,93 g/mol
Boiling Pt: 106 °C (0,7 mmHg)
Melting Pt: 70 °C
Density: 1,745 g/cm³ (20 °C)
CAS Number: 115-29-7
Merck Index: 12,03614

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR