Pipette tips, Microloader
Supplier: Eppendorf
Microloader
612-7933EA 102 EUR
612-7933
Pipette tips, Microloader
Pipette Tips
For filling microinjection capillaries. Ideal for recovering excess solution in the capillary.
Length: 100 mm